Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans,trans-1,5-Bis[4-(trifluoromethyl)phenyl]-1,4-pentadien-3-one 98.0+%, TCI America™
SDP

Catalog No. B44681G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B44681G 1 g
B4468200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B44681G Supplier TCI America Supplier No. B44681G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 103836-71-1
Molecular Formula C19H12F6O
Molecular Weight (g/mol) 370.294
InChI Key OIUBVYQZMUKRRF-YDWXAUTNSA-N
Synonym trans,trans-Bis[4-(trifluoromethyl)benzal]acetone, trans,trans-Bis[4-(trifluoromethyl)benzylidene]acetone, trans,trans-Bis[4-(trifluoromethyl)styryl] Ketone
PubChem CID 5373276
IUPAC Name (1E,4E)-1,5-bis[4-(trifluoromethyl)phenyl]penta-1,4-dien-3-one
SMILES C1=CC(=CC=C1C=CC(=O)C=CC2=CC=C(C=C2)C(F)(F)F)C(F)(F)F

Specifications

Quantity 1 g
Formula Weight 370.29
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material trans,trans-1,5-Bis[4-(trifluoromethyl)phenyl]-1,4-pentadien-3-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.