Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans,trans-2,4-Decadienal, 95%, Tech.

Catalog No. p-3757384
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 25152-84-5 | C10H16O | 152.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25152-84-5
Molecular Formula C10H16O
Molecular Weight (g/mol) 152.24
MDL Number MFCD00007007
InChI Key JZQKTMZYLHNFPL-BLHCBFLLSA-N
Synonym 2e,4e-deca-2,4-dienal, 2,4-decadienal, e,e-2,4-decadienal, trans,trans-2,4-decadienal, 2,4-decadienal, 2e,4e, 2e,4e-decadienal, 2e,4e-2,4-decadienal, 2,4-decadienal, e,e, 2-trans-4-trans-decadienal, ccris 4029
PubChem CID 5283349
IUPAC Name (2E,4E)-deca-2,4-dienal
SMILES CCCCCC=CC=CC=O

Specifications

Color Yellow
Density 0.8500g/mL
Boiling Point 114°C to 116°C (10.0 mmHg)
Flash Point 85°C
Infrared Spectrum Authentic
Assay Percent Range 92% min. (main isomer) (GC)
Packaging Glass bottle
Refractive Index 1.5140 to 1.518
Linear Formula CH3(CH2)4CH=CHCH=CHCHO
Quantity 1 g
Specific Gravity 0.85
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in most organic solvents
Formula Weight 152.24
Percent Purity 95%
Grade Technical
Physical Form Liquid
Chemical Name or Material trans, trans-2, 4-Decadienal
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 246-668-9

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.