Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Transcobalamin II Antibody, mFluor Violet 450 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298341MFV450
Description
Transcobalamin II Polyclonal antibody specifically detects Transcobalamin II in Mouse samples. It is validated for ELISASpecifications
Transcobalamin II | |
Polyclonal | |
ELISA | |
D22S676, D22S750, macrocytic anemia, TC, TC-2, TC2II, TCII, transcobalamin II; macrocytic anemia, transcobalamin IITC II, transcobalamin-2, vitamin B12-binding protein 2 | |
Produced in rabbits immunized with purified, recombinant Mouse Transcobalamin II (Uniprot#: O88968; Met 1-Trp 430) | |
0.1 mL | |
Cardiovascular Biology, Endocrinology, Signal Transduction | |
6948 | |
Store at 4C in the dark. | |
IgG |
ELISA | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction