Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Transcription Factor IIB Monoclonal Antibody (IIB8), QED Bioscience™
SDP

Catalog No. 502278272
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-227-8272 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-227-8272 Supplier Leinco Technologies Inc Supplier No. 70103200UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Transcription Factor IIB Monoclonal antibody specifically detects Transcription Factor IIB in Human, Mouse samples. It is validated for ELISA, Western Blot

Specifications

Antigen Transcription Factor IIB
Applications ELISA, Western Blot
Classification Monoclonal
Clone IIB8
Concentration 0.5-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene GTF2B
Gene Accession No. P62915, Q00403
Gene Alias general transcription factor 2B L homeolog; general transcription factor IIB; general transcription factor TFIIB; GTF2B; gtf2b.L; gtf2b-a; gtf2b-b; LOW QUALITY PROTEIN: transcription initiation factor IIB; MGC137096 protein; RNA polymerase II alpha initiation factor; RNA polymerase II transcription factor IIB; RP11-82K18.1; S300-II; TF2B; tfiib; Transcription initiation factor IIB; Transcription initiation factor IIB-like protein; wu:fc44c11; XELAEV_18023356mg; XLTFIIB; zgc:66258
Host Species Mouse
Immunogen Partially purified recombinant human transcription factor IIB (TFIIB, Promega Corp.).
Purification Method Protein A
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 229906, 2959
Target Species Human, Mouse
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.