Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Transducin alpha Antibody, PerCP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB1203504PCP
Description
Transducin alpha Polyclonal antibody specifically detects Transducin alpha in Mouse, Porcine, Bovine, Sheep samples. It is validated for Western BlotSpecifications
Transducin alpha | |
Polyclonal | |
Western Blot | |
CSNBAD3, GBT1, GNATR, guanine nucleotide binding protein (G protein), alpha transducing activitypolypeptide 1, guanine nucleotide-binding protein G(t) subunit alpha-1, guanine nucleotide-binding protein G(T), alpha-1 subunit, rod-type transducin alpha subunit, Transducin alpha-1 chain, transducin, rod-specific | |
Synthetic Peptide: K(18) L K E D A E K D A R T V K(31) C | |
0.1 mL | |
Signal Transduction, Vision | |
2779 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
PerCP | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Mouse, Pig, Bovine, Sheep | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction