Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Transfer or electro blotting buffer (10X), pH 8.5

Catalog No. AAJ63037K3
Change view
Click to view available options
Quantity:
1L
2L
4L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ63037K3 2L
AAJ63037K2 1L
AAJ63037K7 4L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ63037K3 Supplier Thermo Scientific Chemicals Supplier No. J63037K3
Add to Cart

Used to allows protein transfer from gels to nitrocellulose or PVDF at relatively higher voltages.

Transfer or electro blotting buffer (10X), pH 8.5 is used to allows protein transfer from gels to nitrocellulose or PVDF at relatively higher voltages.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Form Liquid
Quantity 2L
Storage Requirements Ambient temperatures
How can I prepare a 1X buffer solution from the 10X Transfer or electro blotting buffer (10X), pH 8.5?

You can make a 1:10 dilution of 10X Transfer or electro blotting buffer (10X), pH 8.5 with deionized water to create a 1X Transfer or electro blotting buffer. Adjust the pH to 8.5.

What are the concentrations of the components in the Transfer or electro blotting buffer (10X), pH 8.5?

Transfer or electro blotting buffer (10X), pH 8.5 consists of 250 mM Tris and 1.92 M glycine.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.