Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Transfer Ribonucleic Acid, MP Biomedicals™
Click to view available options
Quantity:
1 kU
100 U
5 kU
500 U
Description
- One unit will yield an A260 of 1.0 in 1.0 mL water (1 cm light path).

Specifications
Specifications
Format | Vial (powder) |
For Use With (Application) | DNA and RNA Purification, Enzymes, Substrates, Inhibitors |
Product Type | Transfer Ribonucleic Acid |
Quantity | 100 U |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction