Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Transferrin Antibody (HTF-14), DyLight 405, Novus Biologicals™
SDP

Catalog No. NB500418V Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB500418V 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB500418V Supplier Novus Biologicals Supplier No. NB500418V
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Transferrin Monoclonal specifically detects Transferrin in Human, Porcine, Rabbit, Bovine (Negative), Canine (Negative), Equine (Negative), Sheep (Negative) samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen Transferrin
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone HTF-14
Conjugate DyLight 405
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Gene Alias Beta-1 metal-binding globulin, DKFZp781D0156, EC 3.4.21, PRO1557, PRO2086, serotransferrin, Siderophilin, transferrin
Gene Symbols TF
Host Species Mouse
Immunogen Purified porcine transferrin.
Purification Method Ion exchange chromatography
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cellular Markers, Golgi Apparatus Markers, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 7018
Test Specificity The antibody HTF-14 recognizes an epitope located in the N-terminal domain of human serum transferrin, a 77 kDa single polypeptide chain glycoprotein (member of the iron binding family of proteins). It is synthesised in the liver and consists of two domains each having a high affinity reversible binding site for Fe3+.
Target Species Human, Pig, Rabbit, Bovine (Negative), Canine (Negative), Equine (Negative), Sheep (Negative)
Content And Storage Store at 4C in the dark.
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.