Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Transferrin Antibody (OTI5G2), Alexa Fluor™ 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274583AF350
Description
Transferrin Monoclonal specifically detects Transferrin in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.Specifications
Transferrin | |
Monoclonal | |
Alexa Fluor 350 | |
50 mM sodium borate with 0.05% sodium azide | |
TF | |
Full length recombinant protein of human Transferrin (NP_001054) produced in HEK293T cells. | |
Primary | |
Human | |
IgG1 |
Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
OTI5G2 | |
Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
Beta-1 metal-binding globulin, DKFZp781D0156, EC 3.4.21, PRO1557, PRO2086, serotransferrin, Siderophilin, transferrin | |
Mouse | |
0.1 mL | |
7018 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction