Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Transferrin Antibody (TF/3001), Alexa Fluor™ 532, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP279912AF532
Description
Transferrin Monoclonal specifically detects Transferrin in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Transferrin | |
Monoclonal | |
Alexa Fluor 532 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
TF | |
Recombinant fragment (around aa 311-445) of human Transferrin protein (exact sequence is proprietary) (Uniprot: P02787) | |
0.1 mL | |
Apoptosis, Cancer, Cellular Markers, Golgi Apparatus Markers, Loading Controls, Stem Cell Markers | |
Human | |
IgG1 κ |
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
TF/3001 | |
ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
Beta-1 metal-binding globulin, DKFZp781D0156, EC 3.4.21, PRO1557, PRO2086, serotransferrin, Siderophilin, transferrin | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction