Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Transferrin Chimeric Recombinant Rabbit Monoclonal Antibody (TW4.20)
GREENER_CHOICE

Catalog No. PIMA548035
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA548035 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA548035 Supplier Invitrogen™ Supplier No. MA548035
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Specificity: Binds to the N-terminal domain of human transferrin.

Transferrins (Tf) are an approximately 80kDa blood plasma glycoprotein synthesized by the liver. They are iron binding transport proteins which can bind two Fe(3+) ions in association with the binding of an anion, usually bicarbonate. It is responsible for the transport of iron from sites of absorption and heme degradation to those of storage and utilization. Transferrin is the primary blood iron transport protein and under normal conditions; approximately one-third of total blood transferrin contains bound iron. Measurement of blood transferrin levels can be used as an indicator for blood iron-carrying capacity and abnormalities of iron metabolism such as anaemia, iron overload and haemochromatosis.Serum transferrin may also have a further role in stimulating cell proliferation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Transferrin Chimeric
Applications ELISA, Flow Cytometry, Inhibition Assays, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone TW4.20
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% ProClin 300
Gene Tf
Gene Accession No. P02787
Gene Alias AI266983; beta-1 metal-binding globulin; Cd176; DKFZp781D0156; epididymis secretory sperm binding protein Li 71p; HEL-S-71p; HP; hpx; hypotransferrinemia with hemochromatosis; ICA; Inhibitor of carbonic anhydrase; lactoferrin; liver regeneration-related protein LRRG03; liver transferrin; LOC100146633; PICA; porcine inhibitor of carbonic anhydrase; porcine inhibitor of carbonic anhydrase precursor; PRO1400; PRO1557; PRO2086; Serotransferrin; siderophilin; TF; Tfn; TFQTL1; transferin; Transferrin; transferrin precursor; Trf
Gene Symbols Tf
Host Species Rabbit
Immunogen Iron-free purified human transferrin.
Purification Method Protein A
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7018
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.