Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Transferrin Monoclonal Antibody (D7-D11), Huabio

Catalog No. 502544495
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-4495 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-4495 Supplier HUABIO RESEARCH INC Supplier No. M08095
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Transferrin Monoclonal antibody specifically detects Transferrin in Human samples. It is validated for Western Blot

Specifications

Antigen Transferrin
Applications Western Blot
Classification Monoclonal
Clone D7-D11
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene Tf
Gene Accession No. P02787
Gene Alias AI266983; beta-1 metal-binding globulin; Cd176; DKFZp781D0156; epididymis secretory sperm binding protein Li 71p; HEL-S-71p; HP; hpx; hypotransferrinemia with hemochromatosis; ICA; Inhibitor of carbonic anhydrase; lactoferrin; liver regeneration-related protein LRRG03; liver transferrin; LOC100146633; PICA; porcine inhibitor of carbonic anhydrase; porcine inhibitor of carbonic anhydrase precursor; PRO1400; PRO1557; PRO2086; Serotransferrin; siderophilin; TF; Tfn; TFQTL1; transferin; Transferrin; transferrin precursor; Trf
Host Species Mouse
Immunogen Recombinant protein within human Transferrin aa 20-698.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7018
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.