Learn More
Invitrogen™ Transferrin Monoclonal Antibody (F2H8G6)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MIT0603
Description
MIT0603 has been successfully used in ELISA, immunofluorescence, immunoprecipitation, and radioimmune assay applications, and reacts with human samples. MIT0603 detects Transferrin which has a predicted molecular weight of approximately 67 kDa. MIT0603 was formerly sold as a Seradyn product.
Transferrins (Tf) are an approximately 80kDa blood plasma glycoprotein synthesized by the liver. They are iron binding transport proteins which can bind two Fe(3+) ions in association with the binding of an anion, usually bicarbonate. It is responsible for the transport of iron from sites of absorption and heme degradation to those of storage and utilization. Transferrin is the primary blood iron transport protein and under normal conditions; approximately one-third of total blood transferrin contains bound iron. Measurement of blood transferrin levels can be used as an indicator for blood iron-carrying capacity and abnormalities of iron metabolism such as anaemia, iron overload and haemochromatosis.Serum transferrin may also have a further role in stimulating cell proliferation.
Specifications
Transferrin | |
Monoclonal | |
5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
P02787 | |
Tf | |
Human Transferrin. | |
1 mg | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA, Immunoprecipitation, Radioimmune Assays (RIA), Immunocytochemistry | |
F2H8G6 | |
Unconjugated | |
Tf | |
AI266983; beta-1 metal-binding globulin; Cd176; DKFZp781D0156; epididymis secretory sperm binding protein Li 71p; HEL-S-71p; HP; hpx; hypotransferrinemia with hemochromatosis; ICA; Inhibitor of carbonic anhydrase; lactoferrin; liver regeneration-related protein LRRG03; liver transferrin; LOC100146633; PICA; porcine inhibitor of carbonic anhydrase; porcine inhibitor of carbonic anhydrase precursor; PRO1400; PRO1557; PRO2086; Serotransferrin; siderophilin; TF; Tfn; TFQTL1; transferin; Transferrin; transferrin precursor; Trf | |
Mouse | |
Protein A | |
RUO | |
7018 | |
Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C | |
Liquid |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.