Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Transferrin Native Protein
SDP

Catalog No. NBP196775 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP196775 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP196775 Supplier Novus Biologicals™ Supplier No. NBP196775
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

Native TF. Source: Serum (Accession #: P12346) The Transferrin Native Protein has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation.

Specifications

Accession Number P12346
For Use With (Application) ELISA, Immunohistochemistry, Western Blot
Formulation 0.01 M Sodium Phosphate, 0.25M Sodium Chloride, pH 7.6
Gene ID (Entrez) 7018
Name Transferrin Protein
Quantity 5 mg
Storage Requirements 4°C short term, −20°C long term. Avoid freeze-thaw cycles.
Cross Reactivity Rat
Purity or Quality Grade Multi-step
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.