Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Transferrin R (CD71) Mouse anti-Human , Alexa Fluor 647, Clone: OKT9 (also known as OKT-9; OKT 9), BD Biosciences™

Catalog No. BDB566724 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB566724 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB566724 Supplier BD Biosciences Supplier No. 566724
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Transferrin R (CD71) Monoclonal antibody specifically detects Transferrin R (CD71) in Human samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen Transferrin R (CD71)
Applications Flow Cytometry
Classification Monoclonal
Clone OKT9 (also known as OKT-9; OKT 9)
Conjugate Alexa Fluor 647
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Gene Alias TfR; sTfR; TFRC; Trfr; TfR1; TR; T9; p90
Host Species Mouse
Immunogen Human T-cell acute lymphoblastic leukemia cells
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to Alexa Fluor 647 under optimum conditions, and unreacted Alexa Fluor 647 was removed.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.