Learn More
Invitrogen™ Transferrin Receptor Monoclonal Antibody (ICO-92)
Description
MA1-7657 detects Transferrin receptor from human samples. MA1-7657 has been successfully used in flow cytometry procedures. The MA1-7657 immunogen is a Transferrin receptor on activated T-B-cells.
Specifications
Specifications
| Antigen | Transferrin Receptor |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | ICO-92 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.5% saccharose, 0.2% gelatin and 0.1% sodium azide; pH 7.4 |
| Gene | TFRC |
| Gene Accession No. | P02786 |
| Gene Alias | 2610028K12Rik; AI195355; AI426448; AU015758; CD antigen CD71; CD71; chicken transferrin receptor; E430033M20Rik; I79_000642; IMD46; mammary tumor virus receptor 1; Mtvr1; Mtvr-1; p90; sTfR; T9; TfR; tfR1; TFR2; TFRC; TR; transferrin receptor; transferrin receptor (p90, CD71); transferrin receptor 2; transferrin receptor protein 1; Transferrin receptor protein 1, serum form; Trfr |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.