Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Transgelin 2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
SDP

Catalog No. 501726136 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-6136 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-6136 Supplier Proteintech Group Inc Supplier No. 102342AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody was generated against full length transgenlin 2 protein. It may cross react with other two transgelins based on the sequence similarity.

The protein encoded by this gene is a homolog of the protein transgelin, which is one of the earliest markers of differentiated smooth muscle. The function of this protein has not yet been determined.

Specifications

Antigen Transgelin 2
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.21 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene TAGLN2
Gene Accession No. P37802, Q9WVA4
Gene Alias SM22 beta, SM22beta, TAGLN2, Transgelin 2
Gene Symbols TAGLN2
Host Species Rabbit
Immunogen Transgelin 2 Fusion Protein Ag0337
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21346, 8407
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.