Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
transgelin/SM22 Mouse anti-Human, Mouse, Porcine, Rat, Clone: 2A10C2, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | transgelin/SM22 |
---|---|
Clone | 2A10C2 |
Concentration | 0.7 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6316
![]() ![]() |
Proteintech Group Inc
602131IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
The protein encoded by this gene is a transformation and shape-change sensitive actin cross-linking/gelling protein found in fibroblasts and smooth muscle. Its expression is down-regulated in many cell lines, and this down-regulation may be an early and sensitive marker for the onset of transformation. A functional role of this protein is unclear. Two transcript variants encoding the same protein have been found for this gene.Specifications
transgelin/SM22 | |
0.7 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
SM22, SM22 alpha, SM22a, SM22-a, SM22alpha, Smooth muscle protein 22 alpha, TAGLN, TAGLN1, transgelin, transgelin/SM22 alpha | |
TAGLN | |
IgG1 | |
Protein G | |
Antibody |
2A10C2 | |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human, Mouse, Pig, Rat | |
P31232, P37804, Q01995 | |
21345, 25123, 397021, 6876 | |
transgelin/SM22 Fusion Protein Ag0764 | |
Primary | |
-20°C | |
TAGLN |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title