Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Transgelin/TAGLN/SM22 alpha Antibody (SAIC-33B-129), Alexa Fluor™ 350 , Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP320101AF350
Description
Transgelin/TAGLN/SM22 alpha Monoclonal antibody specifically detects Transgelin/TAGLN/SM22 alpha in Human samples. It is validated for Western Blot, ELISASpecifications
Transgelin/TAGLN/SM22 alpha | |
Monoclonal | |
Alexa Fluor 350 | |
DKFZp686P11128, Protein WS3-10, SM2222 kDa actin-binding protein, SMCCDKFZp686B01212, Smooth muscle protein 22-alpha, TAGLN1, transgelin, transgelin variant 2, WS3-10SM22-alpha | |
Peptide "e;AAEDYGVIK"e;, derived from the actin cross-linking protein Transgelin, conjugated to KLH. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot, ELISA | |
SAIC-33B-129 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
6876 | |
Store at 4°C in the dark. | |
IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction