Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Transglutaminase 1/TGM1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication
$382.00 - $646.00
Specifications
Antigen | Transglutaminase 1/TGM1 |
---|---|
Dilution | Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:2500 - 1:5000 |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
Transglutaminase 1/TGM1 Polyclonal specifically detects Transglutaminase 1/TGM1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Transglutaminase 1/TGM1 | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
Epidermal TGase, ICR2, KTG, LI, LI1EC 2.3.2.13, protein-glutamine gamma-glutamyltransferase K, protein-glutamine-gamma-glutamyltransferase), TG(K), TGASE, TGase-1, TGKTGase K, transglutaminase 1 (K polypeptide epidermal type I, Transglutaminase K, transglutaminase, keratinocyte, transglutaminase-1 | |
TGM1 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:2500 - 1:5000 | |
Polyclonal | |
Rabbit | |
Human | |
P22735 | |
7051 | |
This antibody was developed against a recombinant protein corresponding to amino acids: MMDGPRSDVGRWGGNPLQPPTTPSPEPEPEPDGRSRRGGGRSFWARCCGCCSCRNAADDDWGPEPSDSRGRGSSSGTRRPGSRGSDSRRPVSRGSGVNAA | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title