Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Transglutaminase-2 (TG2) Monoclonal Antibody (7D2), QED Bioscience™

Mouse Monoclonal Antibody
Supplier: Leinco Technologies Inc 90021100UG

This item is not returnable.
View return policy
Description
Transglutaminase-2 (TG2) Monoclonal antibody specifically detects Transglutaminase-2 (TG2) in Guinea Pig, Human samples. It is validated for Immunocytochemistry, Western BlotSpecifications
Transglutaminase-2 (TG2) | |
Monoclonal | |
0.5-1.0 mg/mL | |
0.1M tris with 0.1M glycine, 2% sucrose and no preservative | |
P21980 | |
Mouse | |
Protein A | |
RUO | |
100379228, 7052 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunocytochemistry, Western Blot | |
7D2 | |
Unconjugated | |
TGM2 | |
C polypeptide; C polypeptide, protein-glutamine-gamma-glutamyltransferase; G alpha h; G[a]h; G-ALPHA-h; GNAH; Protein-glutamine gamma-glutamyltransferase 2; protein-glutamine-gamma-glutamyltransferase; RP5-1054A22.2; TG C; TG(C); TG2; TGase C; TGase H; TGase2; TGase-2; TGase-H; TgaseII; TGC; TGM2; Tissue transglutaminase; Tissue type transglutaminase; tissue-type transglutaminase; transglutaminase; transglutaminase 2; transglutaminase 2 (C polypeptide, protein-glutamine-gamma-glutamyltransferase); Transglutaminase 2 C polypeptide; transglutaminase 2, C polypeptide; transglutaminase C; Transglutaminase H; transglutaminase-2; tTG; tTGas | |
Purified TG2 from guinea pig liver (accession no. P21980). | |
100 μg | |
Primary | |
Guinea Pig, Human | |
Antibody | |
IgG3 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction