Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Transglutaminase 2/TGM2 Antibody (7E1), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | Transglutaminase 2/TGM2 |
---|---|
Clone | 7E1 |
Dilution | Western Blot 1:500-1:5000, ELISA |
Applications | Western Blot, ELISA |
Classification | Monoclonal |
Description
Transglutaminase 2/TGM2 Monoclonal antibody specifically detects Transglutaminase 2/TGM2 in Human samples. It is validated for Western Blot, ELISASpecifications
Transglutaminase 2/TGM2 | |
Western Blot 1:500-1:5000, ELISA | |
Monoclonal | |
Purified | |
RUO | |
Human | |
C polypeptide, EC 2.3.2.13, G-ALPHA-h, protein-glutamine gamma-glutamyltransferase 2, protein-glutamine-gamma-glutamyltransferase, TG(C), TG2, TGase C, TGase H, TGase-2, TGase-H, TGCGNAH, Tissue transglutaminase, transglutaminase 2 (C polypeptide, protein-glutamine-gamma-glutamyltransferase), Transglutaminase C, Transglutaminase H, Transglutaminase-2 | |
A synthesized peptide derived from Human Transglutaminase 2/TGM2 [UniProt P21980] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
7E1 | |
Western Blot, ELISA | |
Unconjugated | |
Rabbit | |
Apoptosis, Immune System Diseases, Immunology, Signal Transduction | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
7052 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title