Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Transglutaminase 7/TGM7 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP281763
Description
Transglutaminase 7/TGM7 Polyclonal specifically detects Transglutaminase 7/TGM7 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
Transglutaminase 7/TGM7 | |
Polyclonal | |
Unconjugated | |
PBS | |
TGM7 | |
TGM7 antibody was raised against a 13 amino acid synthetic peptide near the center of human TGM7. The immunogen is located within amino acids 460 - 510 of TGM7. Amino Acid Squence: LESGGLRDQPAQL | |
0.1 mg | |
Cancer | |
116179 | |
Human, Mouse | |
IgG |
Western Blot, ELISA, Immunofluorescence | |
1 mg/mL | |
Western Blot 1 μg/mL, ELISA, Immunocytochemistry/Immunofluorescence 2.5-10 μg/mL | |
protein-glutamine gamma-glutamyltransferase Z, TG(Z), TGase Z, TGase-7, TGMZEC 2.3.2.13, TGZ, transglutaminase 7, Transglutaminase Z, transglutaminase-7 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Transglutaminase 7/TGM7 antibody is predicted to not cross-react with other TGase protein family members. At least two isoforms of Transglutaminase 7/TGM7 are known to exist; this antibody will recognize both isoforms. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction