Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Transglutaminase 7/TGM7 Antibody, DyLight 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP281763UV
This item is not returnable.
View return policy
Description
Transglutaminase 7/TGM7 Polyclonal antibody specifically detects Transglutaminase 7/TGM7 in Human, Mouse samples. It is validated for Western Blot, ELISA, ImmunofluorescenceSpecifications
Transglutaminase 7/TGM7 | |
Polyclonal | |
50mM Sodium Borate | |
Rabbit | |
Peptide affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Western Blot, ELISA, Immunofluorescence | |
DyLight 350 | |
protein-glutamine gamma-glutamyltransferase Z, TG(Z), TGase Z, TGase-7, TGMZEC 2.3.2.13, TGZ, transglutaminase 7, Transglutaminase Z, transglutaminase-7 | |
TGM7 antibody was raised against a 13 amino acid synthetic peptide near the center of human TGM7. The immunogen is located within amino acids 460 - 510 of TGM7. Amino Acid Squence: LESGGLRDQPAQL | |
0.1 mL | |
Cancer | |
116179 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction