Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mirus Bio TransIT-VirusGEN™ Transfection, 150mL
Click to view available options
Quantity:
150 mL
Description
- TransIT-VirusGEN Transfection Reagent is designed to enhance delivery of packaging and transfer plasmids to adherent and suspension HEK 293 cell types to increase recombinant adeno-associated virus (AAV) and lentivirus production.
- Key benefits:
- Performance: Efficient DNA delivery for production of high-titer viral vectors
- Scalability: Efficient production from less than 1 mL to large scale culturesFlexibility: Compatible with different recombinant virus and cell culture systems
- Animal Origin Free: Fully synthetic transfection reagent formulation
Specifications
Specifications
Cell Type | 293-derived Cells |
Format | Bottle |
Promoter | None (Promoterless) |
Transfection Technique | Chemical Transfection |
Quantity | 150 mL |
Sterility | Sterile |
Volume (Metric) | 150 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction