Learn More
Description
TransIT-VirusGEN™ Transfection Reagent is designed to enhance delivery of packaging and transfer vectors to adherent and suspension HEK 293 cell types to increase recombinant adeno-associated virus (AAV) and lentivirus production.
- Reliable – Consistent high virus titer production
- Scalable – Efficient across different formats
- Flexible – Compatible with different recombinant virus and cell culture systems
- Animal Origin Free – High performance with maximum compatibility
Specifications
Specifications
| Content And Storage | Store at -10°C to -30°C |
| Description | Before each use, warm to room temperature and vortex gently. |
| Cell Type | 293-derived Cells |
| Format | Tube |
| Promoter | None (Promoterless) |
| Transfection Technique | Chemical Transfection |
| Product Type | Transfection Reagent |
| Quantity | 1 x 0.75 mL |
| Sterility | Sterile |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.