Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Transketolase Antibody (OTI5H3), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274585MFV500
Description
Transketolase Monoclonal antibody specifically detects Transketolase in Human, Mouse, Rat samples. It is validated for Flow CytometrySpecifications
Transketolase | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Mouse | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Flow Cytometry | |
OTI5H3 | |
Flow Cytometry | |
EC 2.2.1.1, FLJ34765, TK, TKT1, transketolase | |
Full length human recombinant protein of human TKT (NP_001055) produced in HEK293T cell. | |
0.1 mL | |
Stem Cell Markers | |
7086 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction