Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Transketolase (CT) Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS4514R

Description
Transketolase encodes a thiamine-dependent enzyme which plays a role in the channeling of excess sugar phosphates to glycolysis in the pentose phosphate pathway. Multiple alternatively spliced variants, encoding the same protein, have been identified.Specifications
Transketolase (CT) | |
Polyclonal | |
Unconjugated | |
TKT | |
BOS_20935; epididymis luminal protein 107; epididymis secretory protein Li 48; HEL107; HEL-S-48; LOC100350406; P68; TK; Tkt; TKT1; Transketolase; transketolase (Wernicke-Korsakoff syndrome) | |
Rabbit | |
Protein A | |
RUO | |
7086 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
P29401 | |
TKT | |
KLH conjugated synthetic peptide derived from Apple Transketolase (CT). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction