Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Transketolase Mouse anti-Human, Clone: 3E2F3, Proteintech

Mouse Monoclonal Antibody
Supplier: Proteintech Group Inc 660161IG150UL

Description
Transketolase encodes a thiamine-dependent enzyme which plays a role in the channeling of excess sugar phosphates to glycolysis in the pentose phosphate pathway. Multiple alternatively spliced variants, encoding the same protein, have been identified.Specifications
Transketolase | |
Monoclonal | |
1.01 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P29401 | |
TKT | |
Transketolase Fusion Protein Ag16779 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG2b |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
3E2F3 | |
Unconjugated | |
TKT | |
TKT, TKT1, Transketolase | |
Mouse | |
Protein A | |
RUO | |
7086 | |
-20°C | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction