Learn More
Invitrogen™ Transketolase Polyclonal Antibody

Description
Recommended positive controls: Raji, mouse liver, 293T, A431, HeLa, HepG2, human endothelial cells. Predicted reactivity: Mouse (98%), Rat (98%), Xenopus laevis (84%), Pig (99%), Chimpanzee (100%), Bovine (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | Transketolase |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 |
| Gene | TKT |
| Gene Accession No. | P29401, P40142 |
| Gene Alias | epididymis luminal protein 107; epididymis secretory protein Li 48; HEL107; HEL-S-48; p68; TK; TKT; TKT1; Transketolase; transketolase (Wernicke-Korsakoff syndrome) |
| Gene Symbols | TKT |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.