Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Translin Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP176581
Description
Translin Polyclonal specifically detects Translin in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
Translin | |
Polyclonal | |
Unconjugated | |
PBS with 0.02% Sodium Azide | |
BCLF-1, RCHF1, recombination hotspot associated factor, recombination hotspot-binding protein, REHF-1, TBRBP, translin, TRSLN | |
Rabbit | |
Affinity Purified | |
RUO | |
7247 | |
Human, Mouse, Rat | |
IgG |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
1 mg/mL | |
Western Blot 0.5-1 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 5 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 5 μg/mL | |
NP_004613 | |
TSN | |
Antibody was raised against a 20 amino acid synthetic peptide near the carboxy terminus of human Translin. The immunogen is located within the last 50 amino acids of Translin. Amino Acid Squence: NLKNDSLRKRYDGLKYDVKK | |
0.1 mg | |
Primary | |
At least two isoforms of Translin are known to exist; this antibody will detect both. | |
Store at 4°C. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction