Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Globe Scientific Translucent Snap Caps for 16 mm Tubes

Catalog No. 23556035
Change view
Click to view available options
Color:
Blue
Gray
Green
Lavender
Natural
Red
Yellow
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
23-556-035 Natural
23-556-033 Gray
23-556-034 Blue
23-556-036 Green
23-556-037 Lavender
23-556-038 Red
23-556-039 Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 23-556-035 Supplier Globe Scientific Supplier No. 113144C
Add to Cart
Add to Cart

These colorful snap-on closures are an effective way to recap blood drawing tubes, glass test tubes, and plastic culture tubes.

These colorful snap-on closures are precision molded from low-density polyethylene (LDPE) and maintain a tight seal during centrifugation, agitation and refrigeration procedures. They are available in bright translucent colors for sample identification purposes. The caps feature a single thumb tab for easy removal and close nesting in tube racks.

  • Ecomical way to recap glass tubes, regardless of manufacturer
  • Available in seven transparent colors that correspond to Vacutainer™ cap colors: Natural, Blue, Green, Lavender, Red, Gray, and Yellow
  • Caps can be written on with indelible ink for easy sample identification
  • Prevents spills and evaporation by employing a liquid tight seal

Specifications

Closure Size 16 mm
Autoclavable Not Autoclavable
Material LDPE
Color Natural
No. per Pack 1000 Pk.
Sterility Non-sterile
For Use With (Equipment) 16 mm Evacuated Tubes
Disposable Single-use
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.