Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Transmissible Gastroenteritis Virus Monoclonal Antibody (TGEV10-116.3.3), Invitrogen™

Catalog No. PIMA546145
Change view
Click to view available options
Quantity:
1 mg
100 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA546145 1 mg
PIMA546180 100 μg
01-671-813 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. PIMA546145 Supplier Invitrogen™ Supplier No. MA546145
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

MA1-10783 detects Transmissible Gastroenteritis Virus. MA1-10783 has been successfully used in ELISA applications. The MA1-10783 immunogen is TGEV-infected ST cells.

Transmissible gastroenteritis virus or Transmissible gastroenteritis coronavirus is a coronavirus which infects pigs. It is an enveloped, positive-sense, single-stranded RNA virus which enters its host cell by binding to the APN receptor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Transmissible Gastroenteritis Virus
Applications ELISA
Classification Monoclonal
Clone TGEV10-116.3.3
Concentration 1.5 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene Alias TGV
Host Species Mouse
Immunogen TGEV-infected ST cells.
Purification Method Protein A/G
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.