Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Transmissible Gastroenteritis Virus Monoclonal Antibody (TGEV15-886.6)
GREENER_CHOICE

Catalog No. 01671815
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-671-815 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-671-815 Supplier Invitrogen™ Supplier No. MA110785
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA1-10785 detects Transmissible Gastroenteritis Virus. MA1-10785 has been successfully used in ELISA and neutralization applications. The MA1-10785 immunogen is TGEV-infected ST cells.

Transmissible gastroenteritis virus or Transmissible gastroenteritis coronavirus is a coronavirus which infects pigs. It is an enveloped, positive-sense, single-stranded RNA virus which enters its host cell by binding to the APN receptor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Transmissible Gastroenteritis Virus
Applications ELISA
Classification Monoclonal
Clone TGEV15-886.6
Concentration ~1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene Alias TGV
Host Species Mouse
Immunogen TGEV-infected ST cells.
Purification Method Purified
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.