Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Transparent Tape
SCI_ED

Catalog No. S05241 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Transparent tape
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S05241 Transparent tape
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S05241 Supplier Fisher Science Education™ Supplier No. TP5029C/DEL
This item is not returnable. View return policy

Common consumable for use in the science classroom or with kits.

Specifications

Product Type Tape
Color Transparent
For Use With (Application) For science classroom or with kits
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.