Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Transportin 1 Antibody (D45), Allophycocyanin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB6001397APC
Description
Transportin 1 Monoclonal specifically detects Transportin 1 in Human, Mouse, Bovine, Canine, Monkey, Rabbit samples. It is validated for Flow Cytometry, Flow (Intracellular).Specifications
Transportin 1 | |
Monoclonal | |
APC | |
PBS with 0.05% Sodium Azide | |
TNPO1 | |
Human recombinant His-tagged Transportin 1. [UniProt# Q92973] | |
0.1 mL | |
DNA Repair, DNA replication Transcription Translation and Splicing | |
3842 | |
Store at 4C in the dark. | |
IgG1 |
Flow Cytometry | |
D45 | |
Flow Cytometry, Flow (Intracellular) | |
importin beta 2, Importin beta-2, IPO2, karyopherin (importin) beta 2, MIP1Karyopherin beta-2, MIPM9 region interaction protein, transportin 1, transportin-1, TRNKPNB2importin 2 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human, Bovine, Canine, Monkey, Rabbit | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction