Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Transthyretin/Prealbumin Antibody (003), DyLight 405, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290050V
Description
Transthyretin/Prealbumin Monoclonal specifically detects Transthyretin/Prealbumin in Human samples. It is validated for ELISA.Specifications
Transthyretin/Prealbumin | |
Monoclonal | |
DyLight 405 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
3 | |
ELISA | |
ATTR, HsT2651, PALBTBPA, Prealbumin, prealbumin, amyloidosis type I, thyroxine-binding prealbumin, transthyretin | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human Transthyretin/Prealbumin (NP_000362.1; Met 1-Glu 147). | |
0.1 mL | |
Cancer | |
7276 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction