Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Transthyretin/Prealbumin Antibody, DyLight 755, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NBP311028IR
Description
Transthyretin/Prealbumin Polyclonal antibody specifically detects Transthyretin/Prealbumin in Human samples. It is validated for Western Blot, ELISASpecifications
Transthyretin/Prealbumin | |
Polyclonal | |
Western Blot, ELISA | |
ATTR, HsT2651, PALBTBPA, Prealbumin, prealbumin, amyloidosis type I, thyroxine-binding prealbumin, transthyretin | |
Human Transthyretin/Prealbumin | |
0.1 mL | |
Cancer | |
7276 | |
Store at 4°C in the dark. |
Western Blot, ELISA | |
DyLight 755 | |
50mM Sodium Borate | |
Goat | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction