Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trap 101, Tocris Bioscience™

Catalog No. p-4784712
Click to view available options
Quantity:
50 mg
This item is not returnable. View return policy

Catalog No. 25-085-0

Supplier: R&D Systems 2508/50

This item is not returnable. View return policy

Potent and selective NOP antagonist; active in vivo

Potent and selective nociceptin/orphanin FQ (NOP) receptor antagonist (pA 2 = 7.75). Displays selectivity for NOP receptors over classical opioid receptors (pK i values are 8.65, 6.60, 6.14 and < 5 for NOP, μ-, κ-, and δ-opioid receptors respectively). Attenuates motor deficits in a rat model of Parkinson's Disease. Active in vivo.

Chemical Identifiers

CAS 1216621-00-9
Molecular Formula C24H36ClN3O2
Molecular Weight (g/mol) 434.021
InChI Key YEVMLMQAMCKVSC-UHFFFAOYSA-N
Synonym trap 101, trap-101 hydrochloride, 1-1-cyclooctylmethyl-1,2,3,6-tetrahydro-5-hydroxymethyl-4-pyridinyl-3-ethyl-1,3-dihydro-2h-benzimidazol-2-one hydrochloride, 1-1-cyclooctylmethyl-5-hydroxymethyl-1,2,3,6-tetrahydropyridin-4-yl-3-ethyl-1h-benzo d imidazol-2 3h-one hydrochloride, 1-1-cyclooctylmethyl-3-hydroxymethyl-5,6-dihydro-2h-pyridin-4-yl-3-ethyl-1,3-benzodiazol-2-one hydrochloride, trap101-hcl, trap-101 hydrochloride hplc , powder, 1-1-cyclooctylmethyl-1,2,3,6-tetrahydro-5-hydroxymethyl-4-pyridinyl-3-ethyl-1,3-dihydro-2hbenzimidazol-2-one hydrochloride
PubChem CID 56972178
IUPAC Name 1-[1-(cyclooctylmethyl)-5-(hydroxymethyl)-3,6-dihydro-2H-pyridin-4-yl]-3-ethylbenzimidazol-2-one;hydrochloride
SMILES CCN1C2=CC=CC=C2N(C1=O)C3=C(CN(CC3)CC4CCCCCCC4)CO.Cl

Specifications

Quantity 50 mg
Formula Weight 434.02
Percent Purity >98%
Chemical Name or Material Trap 101
Recommended Storage Store at +4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.