Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRAP alpha Antibody (OTI4C7), DyLight 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274591V
Description
TRAP alpha Monoclonal specifically detects TRAP alpha in Human, Mouse, Rat, Canine, Monkey, Primate samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
TRAP alpha | |
Monoclonal | |
DyLight 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
SSR1 | |
Full length recombinant protein of human TRAP alpha (NP_003135) produced in HEK293 cell. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat, Canine, Monkey, Primate | |
IgG1 |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI4C7 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
DKFZp781N23103, FLJ14232, FLJ22100, FLJ23034, FLJ78242, FLJ93042, signal sequence receptor, alpha, SSR alpha subunit, SSR-alpha, translocon-associated protein alpha subunit, translocon-associated protein subunit alpha, TRAP alpha, TRAP-alpha, TRAPASignal sequence receptor subunit alpha | |
Mouse | |
Affinity Purified | |
Cell Cycle and Replication | |
6745.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction