Learn More
TRAP1 Antibody (3H4-2H6), Biotin, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP259343B
Catalog No. NBP259343B
Only null left
Add to Cart
Description
Description
TRAP1 Monoclonal specifically detects TRAP1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
TRAP1
Monoclonal
Biotin
PBS with 0.05% Sodium Azide
TRAP1
Purified recombinant TRAP1
100 μL
Primary
Detects 75kDa.
Store at 4C in the dark.
IgG1 κ
Western Blot, Immunocytochemistry, Immunofluorescence
3H4-2H6
Western Blot, Immunocytochemistry/Immunofluorescence
HSP 75, HSP75TNFR-associated protein 1, HSP90Lheat shock protein 75 kDa, mitochondrial, TNF receptor-associated protein 1, TRAP-1, tumor necrosis factor type 1 receptor associated protein, Tumor necrosis factor type 1 receptor-associated protein
Mouse
Protein G purified
Membrane Trafficking and Chaperones, Stem Cell Markers
10131
Human, Mouse, Rat
Purified
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.