Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRAP1 Antibody (9B6), Novus Biologicals™
SDP

Catalog No. p-5325408 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.05 mg
0.2 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018789 0.025 mg
NBP19749705 0.05 mg
NBP19749702 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NB018789 Supplier Novus Biologicals Supplier No. NBP1974970.025MG
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TRAP1 Monoclonal specifically detects TRAP1 in Human, Mouse samples. It is validated for Western Blot.

Specifications

Antigen TRAP1
Applications Western Blot
Classification Monoclonal
Clone 9B6
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation PBS (pH 7.2) and 50% Glycerol with 0.09% Sodium Azide
Gene Alias HSP 75, HSP75TNFR-associated protein 1, HSP90Lheat shock protein 75 kDa, mitochondrial, TNF receptor-associated protein 1, TRAP-1, tumor necrosis factor type 1 receptor associated protein, Tumor necrosis factor type 1 receptor-associated protein
Gene Symbols TRAP1
Host Species Mouse
Immunogen Recombinant human TRAP1.
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Membrane Trafficking and Chaperones, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 10131
Target Species Mouse, Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.