Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRAP1 Antibody (OTI1H8), PerCP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274592PCP
Description
TRAP1 Monoclonal specifically detects TRAP1 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Flow Cytometry.Specifications
| TRAP1 | |
| Monoclonal | |
| PerCP | |
| PBS with 0.05% Sodium Azide | |
| TRAP1 | |
| Full length recombinant protein of human TRAP1 (NP_057376) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat, Canine, Monkey | |
| IgG2b |
| Flow Cytometry | |
| OTI1H8 | |
| Flow Cytometry | |
| HSP 75, HSP75TNFR-associated protein 1, HSP90Lheat shock protein 75 kDa, mitochondrial, TNF receptor-associated protein 1, TRAP-1, tumor necrosis factor type 1 receptor associated protein, Tumor necrosis factor type 1 receptor-associated protein | |
| Mouse | |
| Affinity Purified | |
| Membrane Trafficking and Chaperones, Stem Cell Markers | |
| 10131.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction