Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRAP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596201
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596201 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596201 Supplier Invitrogen™ Supplier No. PA596201
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: SW620, MCF7, Jurkat, BxPC-3, Mouse kidney, Mouse skeletal muscle, Mouse heart, Mouse brain; Cellular Location: Mitochondrion, Mitochondrion inner membrane, Mitochondrion matrix Immunogen sequence: GLFMREGIVT ATEQEVKEDI AKLLRYESSA LPSGQLTSLS EYASRMRAGT RNIYYLCAPN RHLAEHSPYY EAMKKKDTEV LFCFEQFDEL TLLHLREFDK KKLISVETDI VVDHYKEEKF EDRSPAAECL SEKETEELMA WMRNVLGSRV TNVKVTLRLD THPAMVTVLE MGAARHFLRM QQLAKTQEER AQLLQPTLEI NPRHALIKKL NQLRASEPGL AQLLVDQIYE NAMIAAGLVD DPRAMVGRLN ELLVKALERH.

The 90 kDa heat shock protein (hsp90) family of molecular chaperones is a highly conserved family of proteins that play an important physiological role. Hsp90 is involved in numerous cellular processes but is best known for its association with signal transduction machinery. A recently cloned homolog of hsp90 is the tumor necrosis factor receptor-associated protein (TRAP1). Like hsp90, TRAP1 is found to be associated with numerous proteins involved in diverse actions. Immunofluorescence data has shown TRAP1 to be localized in the mitochondria of mammalian cells. This observation and the fact that TRAP1 is shown to have a mitochondrial targeting presequence strongly implicates TRAP1 as a mitochondrial matrix protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRAP1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.38 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene Trap1
Gene Accession No. Q12931, Q5XHZ0, Q9CQN1
Gene Alias 2410002K23Rik; heat shock protein 75 kDa, mitochondrial; HSP 75; HSP75; HSP90L; testicular tissue protein Li 209; TNF receptor associated protein 1; TNF receptor-associated protein 1; TNFR-associated protein 1; TRAP1; TRAP-1; tumor necrosis factor type 1 receptor associated protein; tumor necrosis factor type 1 receptor-associated protein
Gene Symbols Trap1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 445-704 of human TRAP1 (NP_0573762).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10131, 287069, 68015
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.