Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRAP220 Recombinant Rabbit Monoclonal Antibody (BLR037F)
GREENER_CHOICE

Catalog No. PIMA544269
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA544269 100 μL
PIMA544270 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIMA544269 Supplier Invitrogen™ Supplier No. MA544269
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Epitope retrieval with citrate buffer pH6.0 is recommended for FFPE tissue sections.

Component of the Mediator complex, a coactivator involved in the regulated transcription of nearly all RNA polymerase II-dependent genes. Mediator functions as a bridge to convey information from gene-specific regulatory proteins to the basal RNA polymerase II transcription machinery. Mediator is recruited to promoters by direct interactions with regulatory proteins and serves as a scaffold for the assembly of a functional preinitiation complex with RNA polymerase II and the general transcription factors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRAP220
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone BLR037F
Concentration 100 μg/mL
Conjugate Unconjugated
Formulation BBS with 0.1% BSA and 0.09% sodium azide; pH 8.2
Gene MED1
Gene Accession No. Q15648
Gene Alias Activator-recruited cofactor 205 kDa component; AI480703; ARC205; CRSP1; CRSP200; CRSP210; DRIP205; DRIP230; l11Jus15; MED1; Mediator complex subunit 1; mediator of RNA polymerase II transcription subunit 1; p53 regulatory protein RB18A; Pbp; peroxisome proliferator activated receptor binding protein; peroxisome proliferator-activated receptor-binding protein; Ppar binding protein; PPAR-binding protein; PPARBP; PPARG binding protein; PPARGBP; PPRB; RB18A; RGD1559552; Thyroid hormone receptor-associated protein complex 220 kDa component; thyroid hormone receptor-associated protein complex component TRAP220; thyroid receptor interacting protein 2; thyroid receptor-interacting protein 2; TRAP220; TR-interacting protein 2; TRIP2; TRIP-2; vitamin D receptor-interacting protein 230 kD; vitamin D receptor-interacting protein complex component DRIP205
Gene Symbols MED1
Host Species Rabbit
Immunogen Peptide representing a region between residue 1550 and the C-terminus (residue 1581) of human mediator complex subunit 1.
Purification Method Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5469
Target Species Human
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.