Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRAPPC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5104439
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5104439 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5104439 Supplier Invitrogen™ Supplier No. PA5104439
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total TRAPPC1.

TRAPPC1 plays a role in vesicular transport of proteins to the Golgi apparatus from the endoplasmic reticulum. The encoded protein is a component of the multisubunit transport protein particle (TRAPP) complex. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRAPPC1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene TRAPPC1
Gene Accession No. Q2KMM2, Q5NCF2, Q9Y5R8
Gene Alias BET5; BET5 homolog; BET5-like protein; D11Ertd172e; EGK_08154; melanoma ubiquitous mutated 2; multiple myeloma protein 2; MUM2; MUM-2; novel Sybindin-like family domain-containing protein; trafficking protein particle complex 1; trafficking protein particle complex subunit 1; TRAPPC1; zgc:100969
Gene Symbols TRAPPC1
Host Species Rabbit
Immunogen A synthesized peptide derived from human TRAPPC1(Accession Q9Y5R8), corresponding to amino acid residues M1-F48.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 245828, 287427, 58485
Target Species Human, Mouse, Rat, Monkey
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.