Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRAPPC10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA593214
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA593214 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA593214 Supplier Invitrogen™ Supplier No. PA593214
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MDASEEPLPP VIYTMENKPI VTCAGDQNLF TSVYPTLSQQ LPREPMEWRR SYGRAPKMIH LESNFVQFKE ELLPKEGNKA LLTFPFLHIY WTECCDTEVY KATVKDDLTK WQNVLKAHSS VDWLIVIVEN DAKKKNKTNI LPRTSIVDKI RNDFCNKQSD RCVVLSDPLK DSSRTQESWN AFLTKLRTLL LMSFTKNLGK FEDDMRTLRE KRTEPGWSFC EYFMVQEELA FVFEMLQQFE DALVQYDELD ALFSQYVVNF; Positive Samples: Mouse kidney, Rat lung; Cellular Location: Golgi apparatus, cis-Golgi network.

TRAPPC10, also known as TMEM1 (transmembrane protein 1), EHOC1 (epilepsy holoprosencephaly candidate 1 protein) or GT334, is a widely expressed 1,259 amino acid protein that may function in vesicular transport. Despite its name, TRAPPC10 does not contain transmembrane domains. It is the human ortholog of the yeast Trs130 protein and its structure and function appears to be conserved. Localizing to the cis-Golgi apparatus, TRAPPC10 is believed to be involved in transport from the endoplasmic reticulum (ER) to the Golgi functioning as a component of the multisubunit transport protein particle (TRAPP) complex. Mutations in the gene encoding TRAPPC10 may be involved in autoimmune polyglandular disease type 1 or Unverricht-Lundborg disease, an autosomal recessive type of progressive myoclonic epilepsy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRAPPC10
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene TRAPPC10
Gene Accession No. Q3TLI0
Gene Alias B230307C21Rik; EHOC1; EHOC-1; epilepsy holoprosencephaly candidate 1 protein; epilepsy holoprosencephaly candidate-1 protein; Gm870; GT334; Protein GT334; TMEM1; trafficking protein particle complex 10; trafficking protein particle complex subunit 10; trafficking protein particle complex subunit 10; LOW QUALITY PROTEIN: trafficking protein particle complex subunit 10; trafficking protein particle complex subunit 130; trafficking protein particle complex subunit TMEM1; transmembrane protein 1; transport protein particle subunit TMEM1; TRAPP 130 kDa subunit; TRAPP subunit TMEM1; trappc10; TRS130; TRS30; wu:fc21a02
Gene Symbols TRAPPC10
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-260 of human TRAPPC10 (NP_0032653).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 216131, 309678
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.