Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRAPPC11 Rabbit anti-Human, Polyclonal, R&D Systems™
SDP

Catalog No. NB125021 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB125021 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB125021 Supplier R&D Systems Supplier No. NBP310060100UL

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Rabbit Polyclonal Antibody

TRAPPC11 Polyclonal specifically detects TRAPPC11 in Human samples. It is validated for Western Blot.

Specifications

Antigen TRAPPC11
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml
Formulation PBS buffer, 2% sucrose
Gene Alias foigr, C4orf41, chromosome 4 open reading frame 41, gry, trafficking protein particle complex 11, TRAPPC11
Host Species Rabbit
Immunogen The immunogen is a synthetic peptide directed towards the N terminal region of human C4ORF41 (NP_068761.4). Peptide sequence KGILKTGWMNKHLNLVPALVVVFYELDWDEPQWKEKQSECATRVEIVRQS
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 60684
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Product Type Antibody
Form Purified
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.