Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRAPPC13 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558098
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558098 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558098 Supplier Invitrogen™ Supplier No. PA558098
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LGETFSSYIS VHNDSNQVVK DILVKADLQT SSQRLNLSAS NAAVAELKPD CCIDDVIHHE VKEIGTHILV CAVSY Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

The function of C5orf44 has not been widely studied, and is yet to be fully elucidated.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRAPPC13
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Trappc13
Gene Accession No. A5PLN9
Gene Alias 2410002O22Rik; 2610524F24Rik; AI451892; AU017229; C5orf44; RGD1306583; trafficking protein particle complex 13; trafficking protein particle complex subunit 13; TRAPPC13; Trs65-related; UPF0533 protein C5orf44; UPF0533 protein C5orf44 homolog
Gene Symbols Trappc13
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TRAPPC13. Recombinant protein control fragment (Product #RP-96832).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80006
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.