Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRAPPC3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101931
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101931 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101931 Supplier Invitrogen™ Supplier No. PA5101931
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total TRAPPC3.

TRAPPC3 is a component of the TRAPP complex, which is involved in tethering of transport vesicles to the cis-Golgi membrane.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRAPPC3
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene TRAPPC3
Gene Accession No. O43617, O55013, Q5U1Z2
Gene Alias 1110058K12Rik; AI835942; AU019197; Bet3; BET3 homolog; CDABP0066; trafficking protein particle complex 3; trafficking protein particle complex subunit 3; TRAPPC3
Gene Symbols TRAPPC3
Host Species Rabbit
Immunogen A synthesized peptide derived from human TRAPPC3(Accession O43617), corresponding to amino acid residues K12-R62.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27095, 27096, 362599
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.